N-(4-Methoxyphenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide

Supplier: Matrix Scientific

063729-500MG
101926-266EA 364.62 USD
101926-266
N-(4-Methoxyphenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide
N-(4-Methoxyphenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide

Hazard: Irritant

Formula: C₁₅H₁₈N₄O₂S
MDL Number: MFCD15146509
CAS Number: 1217862-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR