N-(2-Fluorophenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide

Supplier: Matrix Scientific

063728-500MG
101926-264EA 364.62 USD
101926-264
N-(2-Fluorophenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide
N-(2-Fluorophenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₅FN₄OS
MDL Number: MFCD15146508
CAS Number: 1217862-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR