N-(4-Methylphenyl)-5-piperidin-3-yl-1,3,4-thiadiazole-2-carboxamide

Supplier: Matrix Scientific

063725-500MG
101926-258EA 364.62 USD
101926-258
N-(4-Methylphenyl)-5-piperidin-3-yl-1,3,4-thiadiazole-2-carboxamide
N-(4-Methylphenyl)-5-piperidin-3-yl-1,3,4-thiadiazole-2-carboxamide

Hazard: Irritant

Formula: C₁₅H₁₈N₄OS
MDL Number: MFCD15146505
CAS Number: 1217862-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR