5-(Aminomethyl)-N-phenyl-1,3,4-thiadiazole-2-carboxamide

Supplier: Matrix Scientific

063724-500MG
101926-256EA 484.62 USD
101926-256
5-(Aminomethyl)-N-phenyl-1,3,4-thiadiazole-2-carboxamide
5-(Aminomethyl)-N-phenyl-1,3,4-thiadiazole-2-carboxamide

Hazard: Irritant

Formula: C₁₀H₁₀N₄OS
MDL Number: MFCD15146504
CAS Number: 1217862-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR