5-(Aminomethyl)-N-(2,4-difluorophenyl)-1,3,4-thiadiazole-2-carboxamide

Supplier: Matrix Scientific

063723-500MG
101926-254EA 484.62 USD
101926-254
5-(Aminomethyl)-N-(2,4-difluorophenyl)-1,3,4-thiadiazole-2-carboxamide
5-(Aminomethyl)-N-(2,4-difluorophenyl)-1,3,4-thiadiazole-2-carboxamide

Hazard: Irritant

Formula: C₁₀H₈F₂N₄OS
MDL Number: MFCD15146503
CAS Number: 1217862-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR