N-(3-Chlorophenyl)-5-piperidin-3-yl-1,3,4-thiadiazole-2-carboxamide

Supplier: Matrix Scientific

063721-500MG
101926-250EA 243.08 USD
101926-250
N-(3-Chlorophenyl)-5-piperidin-3-yl-1,3,4-thiadiazole-2-carboxamide
N-(3-Chlorophenyl)-5-piperidin-3-yl-1,3,4-thiadiazole-2-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₅ClN₄OS
MDL Number: MFCD15146501
CAS Number: 1217862-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR