N-(2,4-Difluorophenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide

Supplier: Matrix Scientific

063720-500MG
101926-248EA 243.08 USD
101926-248
N-(2,4-Difluorophenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide
N-(2,4-Difluorophenyl)-5-piperidin-4-yl-1,3,4-thiadiazole-2-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₄F₂N₄OS
MDL Number: MFCD15146500
CAS Number: 1217862-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR