N-(4-Chlorophenyl)-5-pyrrolidin-2-yl-1,3,4-thiadiazole-2-carboxamide

Supplier: Matrix Scientific

063719-500MG
101926-246EA 243.08 USD
101926-246
N-(4-Chlorophenyl)-5-pyrrolidin-2-yl-1,3,4-thiadiazole-2-carboxamide
N-(4-Chlorophenyl)-5-pyrrolidin-2-yl-1,3,4-thiadiazole-2-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₃ClN₄OS
MDL Number: MFCD15146499
CAS Number: 1217862-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR