4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
Supplier: Matrix Scientific
063718-500MG
101926-244EA
484.62
USD
101926-244
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
Hazard: Irritant
Formula:
C₂H₆N₆S |
MDL Number:
MFCD00003098 CAS Number: 1750-12-5 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...