Ethyl-1-(cyanomethyl)-1H-pyrazole-3-carboxylate

Supplier: Matrix Scientific

063717-500MG
101926-242EA 484.62 USD
101926-242
Ethyl-1-(cyanomethyl)-1H-pyrazole-3-carboxylate
Ethyl-1-(cyanomethyl)-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₈H₉N₃O₂
MDL Number: MFCD15146498
CAS Number: 1217863-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR