Ethyl-1-(cyanomethyl)-1H-pyrazole-5-carboxylate

Supplier: Matrix Scientific

063716-500MG
101926-240EA 484.62 USD
101926-240
Ethyl-1-(cyanomethyl)-1H-pyrazole-5-carboxylate
Ethyl-1-(cyanomethyl)-1H-pyrazole-5-carboxylate

Hazard: Irritant

Formula: C₈H₉N₃O₂
MDL Number: MFCD15146497
CAS Number: 1217862-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR