1-(3-Hydroxy-1-adamantyl)azepan-2-one

Supplier: Matrix Scientific

063689-500MG
101926-208EA 364.62 USD
101926-208
1-(3-Hydroxy-1-adamantyl)azepan-2-one
1-(3-Hydroxy-1-adamantyl)azepan-2-one

Hazard: Irritant

Formula: C₁₆H₂₅NO₂
MDL Number: MFCD09743468
CAS Number: 1134899-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR