1-(1-Adamantyl)pyrrolidin-2-one

Supplier: Matrix Scientific

063688-500MG
101926-206EA 221.54 USD
101926-206
1-(1-Adamantyl)pyrrolidin-2-one
1-(1-Adamantyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD01749832
CAS Number: 3716-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR