5,7-Dimethyladamantane-1,3-dicarboxylic acid

Supplier: Matrix Scientific

063683-500MG
101926-196EA 183.08 USD
101926-196
5,7-Dimethyladamantane-1,3-dicarboxylic acid
5,7-Dimethyladamantane-1,3-dicarboxylic acid

Hazard: Irritant

Formula: C₁₄H₂₀O₄
MDL Number: MFCD01825788
CAS Number: 13928-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR