4-tert-Butyl-2-nitrophenol

Supplier: Matrix Scientific

Synonyms: 4-(tert-Butyl)-2-nitrophenol

063678-500MG
101926-186EA 181.7 USD
101926-186
4-tert-Butyl-2-nitrophenol
4-tert-Butyl-2-nitrophenol

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MW: 195.2151 g/mol
MDL Number: MFCD00191888
CAS Number: 3279-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR