2-[(1-Adamantyloxy)methyl]oxirane

Supplier: Matrix Scientific

063667-500MG
101926-164EA 243.08 USD
101926-164
2-[(1-Adamantyloxy)methyl]oxirane
2-[(1-Adamantyloxy)methyl]oxirane

Hazard: Irritant

Formula: C₁₃H₂₀O₂
MDL Number: MFCD09036003
CAS Number: 130187-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR