1,4-Dimethyladamantane ≥95%
Supplier: Matrix Scientific
063661-500MG
101926-152EA
646.15
USD
101926-152
1,4-Dimethyladamantane ≥95%
1,4-Dimethyladamantane
Hazard: Irritant
Formula:
C₁₂H₂₀ |
MDL Number:
MFCD09743483 CAS Number: 16267-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...