Isopropyl adamantane-1-carboxylate

Supplier: Matrix Scientific

063659-500MG
101926-148EA 183.08 USD
101926-148
Isopropyl adamantane-1-carboxylate
Isopropyl adamantane-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₂O₂
MDL Number: MFCD00193926
CAS Number: 24556-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR