Propyl adamantane-1-carboxylate

Supplier: Matrix Scientific

063658-500MG
101926-146EA 183.08 USD
101926-146
Propyl adamantane-1-carboxylate
Propyl adamantane-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₂O₂
MDL Number: MFCD04346161
CAS Number: 24556-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR