(2,4-Dimethyl-3,5-dinitrophenyl)amine

Supplier: Matrix Scientific

063643-500MG
101926-116EA 243.08 USD
101926-116
(2,4-Dimethyl-3,5-dinitrophenyl)amine
(2,4-Dimethyl-3,5-dinitrophenyl)amine

Hazard: Irritant

Formula: C₈H₉N₃O₄
MDL Number: MFCD06797875
CAS Number: 500729-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR