4-Bromo-3-nitrophenyl thiocyanate

Supplier: Matrix Scientific

063638-500MG
101926-106EA 243.08 USD
101926-106
4-Bromo-3-nitrophenyl thiocyanate
4-Bromo-3-nitrophenyl thiocyanate

Hazard: Irritant

Formula: C₇H₃BrN₂O₂S
MDL Number: MFCD09036136
CAS Number: 408328-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR