5,6-Dibromo-1,2-dihydroacenaphthylene

Supplier: Matrix Scientific

063627-500MG
101926-086EA 176.92 USD
101926-086
5,6-Dibromo-1,2-dihydroacenaphthylene
5,6-Dibromo-1,2-dihydroacenaphthylene

Hazard: Irritant

Formula: C₁₂H₈Br₂
MDL Number: MFCD00191989
CAS Number: 19190-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR