5,6-Dinitro-2,3-dihydro-1,4-benzodioxine

Supplier: Matrix Scientific

063625-500MG
101926-082EA 243.08 USD
101926-082
5,6-Dinitro-2,3-dihydro-1,4-benzodioxine
5,6-Dinitro-2,3-dihydro-1,4-benzodioxine

Hazard: Irritant

Formula: C₈H₆N₂O₆
MDL Number: MFCD01116989
CAS Number: 59820-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR