6H-Indolo[2,3-b]quinoxaline

Supplier: Matrix Scientific

063624-500MG
101926-080EA 281.54 USD
101926-080
6H-Indolo[2,3-b]quinoxaline
6H-Indolo[2,3-b]quinoxaline

Hazard: Irritant

Formula: C₁₄H₉N₃
MDL Number: MFCD00068754
CAS Number: 84055-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR