4-(1H-1,2,3-Benzotriazol-1-ylmethyl)benzohydrazide

Supplier: Matrix Scientific

063622-500MG
101926-076EA 364.62 USD
101926-076
4-(1H-1,2,3-Benzotriazol-1-ylmethyl)benzohydrazide
4-(1H-1,2,3-Benzotriazol-1-ylmethyl)benzohydrazide

Hazard: Irritant

Formula: C₁₄H₁₃N₅O
MDL Number: MFCD09752670
CAS Number: 870766-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR