1-Adamantylcyanamide

Supplier: Matrix Scientific

063613-500MG
101926-060EA 364.62 USD
101926-060
1-Adamantylcyanamide
1-Adamantylcyanamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂
MDL Number: MFCD01321096
CAS Number: 15784-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR