3-Hydroxyadamantane-1-carbohydrazide

Supplier: Matrix Scientific

063608-500MG
101926-050EA 444.62 USD
101926-050
3-Hydroxyadamantane-1-carbohydrazide
3-Hydroxyadamantane-1-carbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₈N₂O₂
Melting Pt: 173 °C
MDL Number: MFCD02208148
CAS Number: 68435-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR