Bicyclo[3.3.1]nonane-2,6-dione

Supplier: Matrix Scientific

063606-500MG
101926-048EA 364.62 USD
101926-048
Bicyclo[3.3.1]nonane-2,6-dione
Bicyclo[3.3.1]nonane-2,6-dione

Hazard: Irritant

Formula: C₉H₁₂O₂
MDL Number: MFCD00153926
CAS Number: 16473-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR