N,N-Diethyl-4-[(E)-2-nitrovinyl]aniline

Supplier: Matrix Scientific

063603-500MG
101926-042EA 243.08 USD
101926-042
N,N-Diethyl-4-[(E)-2-nitrovinyl]aniline
N,N-Diethyl-4-[(E)-2-nitrovinyl]aniline

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD00544872
CAS Number: 147764-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR