N-(4-Oxo-1-adamantyl)acetamide

Supplier: Matrix Scientific

063600-500MG
101926-036EA 335.38 USD
101926-036
N-(4-Oxo-1-adamantyl)acetamide
N-(4-Oxo-1-adamantyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD01825779
CAS Number: 16790-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR