3,5,7-Trimethyladamantane-1-carboxylic acid

Supplier: Matrix Scientific

063596-500MG
101926-028EA 243.08 USD
101926-028
3,5,7-Trimethyladamantane-1-carboxylic acid
3,5,7-Trimethyladamantane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₂₂O₂
MDL Number: MFCD00187581
CAS Number: 15291-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR