1-Bromo-3-(1-bromoethyl)adamantane
Supplier: Matrix Scientific
063594-500MG
101926-024EA
183.08
USD
101926-024
1-Bromo-3-(1-bromoethyl)adamantane
1-Bromo-3-(1-bromoethyl)adamantane
Hazard: Irritant
Formula:
C₁₂H₁₈Br₂ |
MDL Number:
MFCD00153997 CAS Number: 37845-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...