4-Methyl-1,2,3-thiadiazole-5-carbohydrazide

Supplier: Matrix Scientific

063592-500MG
101926-020EA 484.62 USD
101926-020
4-Methyl-1,2,3-thiadiazole-5-carbohydrazide
4-Methyl-1,2,3-thiadiazole-5-carbohydrazide

Hazard: Irritant

Formula: C₄H₆N₄OS
MDL Number: MFCD00052212
CAS Number: 75423-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR