4'-Aminobiphenyl-4-ol

Supplier: Matrix Scientific

Synonyms: 4'-Amino-[1,1'-biphenyl]-4-ol

063591-500MG
101926-018EA 150.77 USD
101926-018
4'-Aminobiphenyl-4-ol
4'-Aminobiphenyl-4-ol

Hazard: Irritant

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
MDL Number: MFCD00463439
CAS Number: 1204-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR