(+)-Taxifolin

Supplier: Matrix Scientific

Synonyms: (2R,3R)-2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxychroman-4-one

063582-500MG
101926-002EA 183.08 USD
101926-002
(+)-Taxifolin
(+)-Taxifolin

Hazard: Irritant

Formula: C₁₅H₁₂O₇
MW: 304.2516 g/mol
MDL Number: MFCD00006845
CAS Number: 480-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR