(4Z)-3-Methyl-1-phenyl-1H-pyrazole-4,5-dione-4-oxime

Supplier: Matrix Scientific

063581-500MG
101926-000EA 146.15 USD
101926-000
(4Z)-3-Methyl-1-phenyl-1H-pyrazole-4,5-dione-4-oxime
(4Z)-3-Methyl-1-phenyl-1H-pyrazole-4,5-dione-4-oxime

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
MDL Number: MFCD00448519
CAS Number: 1080-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR