(2Z)-4-[(2-Ethoxyphenyl)amino]-4-oxobut-2-enoic acid

Supplier: Matrix Scientific

063580-500MG
101925-998EA 183.08 USD
101925-998
(2Z)-4-[(2-Ethoxyphenyl)amino]-4-oxobut-2-enoic acid
(2Z)-4-[(2-Ethoxyphenyl)amino]-4-oxobut-2-enoic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
MDL Number: MFCD00134740
CAS Number: 1089327-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR