Bis(3-iodophenyl)methanone

Supplier: Matrix Scientific

063579-500MG
101925-996EA 146.15 USD
101925-996
Bis(3-iodophenyl)methanone
Bis(3-iodophenyl)methanone

Hazard: Irritant

Formula: C₁₃H₈I₂O
MDL Number: MFCD00227471
CAS Number: 25186-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR