N-(1-Bromo-2-naphthyl)acetamide

Supplier: Matrix Scientific

063575-500MG
101925-988EA 146.15 USD
101925-988
N-(1-Bromo-2-naphthyl)acetamide
N-(1-Bromo-2-naphthyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₀BrNO
MDL Number: MFCD00226708
CAS Number: 7597-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR