2-[2-(5-Mercapto-1H-1,2,4-triazol-3-yl)ethyl]-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

063573-500MG
101925-984EA 364.62 USD
101925-984
2-[2-(5-Mercapto-1H-1,2,4-triazol-3-yl)ethyl]-1H-isoindole-1,3(2H)-dione
2-[2-(5-Mercapto-1H-1,2,4-triazol-3-yl)ethyl]-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₂H₁₀N₄O₂S
MDL Number: MFCD02648743
CAS Number: 362004-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR