1,4-Dimethyl-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-sulfonyl chloride
Supplier: Matrix Scientific
063572-500MG
101925-982EA
335.38
USD
101925-982
1,4-Dimethyl-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-sulfonyl chloride
1,4-Dimethyl-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-sulfonyl chloride
Hazard: Irritant
Formula:
C₁₀H₉ClN₂O₄S Melting Pt: 250…252 °C |
MDL Number:
MFCD15146486 CAS Number: 841275-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...