(1E)-N,N'-Diphenylethanimidamide

Supplier: Matrix Scientific

063571-500MG
101925-980EA 183.08 USD
101925-980
(1E)-N,N'-Diphenylethanimidamide
(1E)-N,N'-Diphenylethanimidamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂
MDL Number: MFCD00137802
CAS Number: 1452517-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR