1-Amino-2-methyl-9,10-anthraquinone

Supplier: Matrix Scientific

063570-500MG
101925-978EA 146.15 USD
101925-978
1-Amino-2-methyl-9,10-anthraquinone
1-Amino-2-methyl-9,10-anthraquinone

Hazard: Irritant

Formula: C₁₅H₁₁NO₂
MDL Number: MFCD00001220
CAS Number: 82-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR