Pentachlorophenyl-N-[(benzyloxy)carbonyl]glycinate

Supplier: Matrix Scientific

063569-500MG
101925-976EA 146.15 USD
101925-976
Pentachlorophenyl-N-[(benzyloxy)carbonyl]glycinate
Pentachlorophenyl-N-[(benzyloxy)carbonyl]glycinate

Hazard: Irritant

Formula: C₁₆H₁₀Cl₅NO₄
MW: 457.53 g/mol
Melting Pt: 133…134 °C
MDL Number: MFCD00371500
CAS Number: 4824-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR