N,N'-Dimethylethanebis(thioamide)
Supplier: Matrix Scientific
063567-500MG
101925-972EA
146.15
USD
101925-972
N,N'-Dimethylethanebis(thioamide)
N,N'-Dimethylethanebis(thioamide)
Hazard: Irritant
Formula:
C₄H₈N₂S₂ |
MDL Number:
MFCD00022131 CAS Number: 120-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...