(2E)-3-Mesityl-1-phenylprop-2-en-1-one
Supplier: Matrix Scientific
063564-500MG
101925-966EA
243.08
USD
101925-966
(2E)-3-Mesityl-1-phenylprop-2-en-1-one
(2E)-3-Mesityl-1-phenylprop-2-en-1-one
Hazard: Irritant
Formula:
C₁₈H₁₈O |
MDL Number:
MFCD00429722 CAS Number: 87172-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...