Ethyl-2-(acetylamino)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

063563-500MG
101925-964EA 243.08 USD
101925-964
Ethyl-2-(acetylamino)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Ethyl-2-(acetylamino)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₇NO₃S
MDL Number: MFCD00087275
CAS Number: 5919-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR