4-(Dichloromethyl)-2,3,5,6-tetramethylbenzaldehyde

Supplier: Matrix Scientific

063562-500MG
101925-962EA 364.62 USD
101925-962
4-(Dichloromethyl)-2,3,5,6-tetramethylbenzaldehyde
4-(Dichloromethyl)-2,3,5,6-tetramethylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₄Cl₂O
MDL Number: MFCD00447911
CAS Number: 148990-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR