3-(Hydroxymethyl)-2,4,6-trimethylbenzaldehyde

Supplier: Matrix Scientific

063561-500MG
101925-960EA 243.08 USD
101925-960
3-(Hydroxymethyl)-2,4,6-trimethylbenzaldehyde
3-(Hydroxymethyl)-2,4,6-trimethylbenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD00297149
CAS Number: 137380-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR