3-(Dichloromethyl)-1,2,4,5-tetramethylbenzene

Supplier: Matrix Scientific

063559-500MG
101925-956EA 243.08 USD
101925-956
3-(Dichloromethyl)-1,2,4,5-tetramethylbenzene
3-(Dichloromethyl)-1,2,4,5-tetramethylbenzene

Hazard: Irritant

Formula: C₁₁H₁₄Cl₂
MDL Number: MFCD00474864
CAS Number: 135962-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR